Learn More
Med Vet International Miltex Von Graefe Eye Dressing Forceps, 2.75in, Curved, Serrated, German, 18-818
Catalog No. 502830494
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Von Graefe Eye Dressing Forceps. Graefe Dressing Forceps is an extremely useful, multipurpose device, which allows surgeons for grasping, holding, and carrying wound dressings and sutures during surgeries. They are made from premium-grade German stainless steel. Specifications: 2.75in, Curved, Serrated, 18-818
Catalog No. 50-283-0494 Supplier Med Vet International Supplier No. MIL18818
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Von Graefe Eye Dressing Forceps. Graefe Dressing Forceps is an extremely useful, multipurpose device, which allows surgeons for grasping, holding, and carrying wound dressings and sutures during surgeries. They are made from premium-grade German stainless steel. Specifications: 2.75in, Curved, Serrated, 18-818