Learn More
Med Vet International Miltex Padgett Wire Twisting Forceps, 5-5/8in Length, 13mm Jaw Length, German, Pm-914
Catalog No. 502830089
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Wire Twisting Forceps. Wire Twisting Forceps are specially designed forceps required to manipulate and twist off wire after it has been secured. They are made from premium-grade German stainless steel. Specifications: 5-5/8in Length, 13mm Jaw Length, PM-914
Catalog No. 50-283-0089 Supplier Med Vet International Supplier No. MILPM914
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Wire Twisting Forceps. Wire Twisting Forceps are specially designed forceps required to manipulate and twist off wire after it has been secured. They are made from premium-grade German stainless steel. Specifications: 5-5/8in Length, 13mm Jaw Length, PM-914