Learn More
Fine Science Tools FORCEP WITH TEETH ON END
SDP
Catalog No. NC9743190
Encompass
Tip Shape: Straight/ Tips: Fine Teeth/ Tip Dimensions: 4.5 x 1.25mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm
Catalog No. NC9743190 Supplier Fine Science Tools Supplier No. 1103012
Tip Shape: Straight/ Tips: Fine Teeth/ Tip Dimensions: 4.5 x 1.25mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm