Learn More
Med Vet International Kern Bone Holding Forceps, 6.25 in., Von Klaus German Surgical Steel
Catalog No. 502091885
Encompass_Preferred
Kern bone holding forceps features a ratchet and a curved bottom for additional grip and control. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1885 Supplier Med Vet International Supplier No. VK1353516
Kern bone holding forceps features a ratchet and a curved bottom for additional grip and control. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.