Learn More
Med Vet International Ruskin-Liston Bone Cutting Forceps, 7.125 in, Angled on Flat, Von Klaus German Surgical Steel
Catalog No. 502091888
Encompass_Preferred
Features a double action, wide jaws, and they are angled on flat. Ruskin-liston forceps are generally used with small and delicate bones. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1888 Supplier Med Vet International Supplier No. VK1335718
Features a double action, wide jaws, and they are angled on flat. Ruskin-liston forceps are generally used with small and delicate bones. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.