Learn More
Med Vet International Dressing Forceps, 4.5 in., Standard Pattern, Serrated Tips, Von Klaus German Surgical Steel
Catalog No. 502091893
Encompass_Preferred
These dressing forceps have a standard pattern and serrated tips. Dressing forceps are commonly used for holding cotton and gauze during surgical procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1893 Supplier Med Vet International Supplier No. VK1060011
These dressing forceps have a standard pattern and serrated tips. Dressing forceps are commonly used for holding cotton and gauze during surgical procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.