Learn More
Med Vet International Foerster Sponge Forceps, 9.5in, Straight, Serrated Jaws, German Grade, Von Klaus
Catalog No. 502091899
Encompass_Preferred
9.5in. Foerster Sponge Forceps is a German grade surgical instrument. They feature serrated jaws and are strasight. They are generally used in surgical procedures to hold gauze. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1899 Supplier Med Vet International Supplier No. VK1195024
9.5in. Foerster Sponge Forceps is a German grade surgical instrument. They feature serrated jaws and are strasight. They are generally used in surgical procedures to hold gauze. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.