Learn More
Med Vet International 5-in Von Klaus Halsted Mosquito Forceps, Straight, Standard Pattern, German Surgical Steel
Catalog No. 502091901
Encompass_Preferred
5" Halsted Mosquito Forceps is a German sugical steel instrument and are primarily used as a hemostat for small blood vessles. They are straight with a standard pattern and are very similar to Hartmann Mosquito Forceps. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1901 Supplier Med Vet International Supplier No. VK1154412
5" Halsted Mosquito Forceps is a German sugical steel instrument and are primarily used as a hemostat for small blood vessles. They are straight with a standard pattern and are very similar to Hartmann Mosquito Forceps. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.