Learn More
Med Vet International 6.25-in Von Klaus Kelly-Rankin Forceps, Curved, German Surgical Steel
Catalog No. 502091909
Encompass_Preferred
6.25-in. Kelly-Rankin forceps are curved and made of German surgical steel. They are typically used for clamping large blood vessels or manipulating heavy tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1909 Supplier Med Vet International Supplier No. VK1157116
6.25-in. Kelly-Rankin forceps are curved and made of German surgical steel. They are typically used for clamping large blood vessels or manipulating heavy tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.