Learn More
Med Vet International Iris Thumb Forceps, 4in Dressing Forceps, Serrated Tips, German Grade, Von Klaus
Catalog No. 502091918
Encompass_Preferred
4' Iris Thumb Forceps is a German grade surgical instrument. They are straight and have serrated tips. Iris Thumb Forceps are great for holding dressings during ophthalmic procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1918 Supplier Med Vet International Supplier No. VK1063010
4' Iris Thumb Forceps is a German grade surgical instrument. They are straight and have serrated tips. Iris Thumb Forceps are great for holding dressings during ophthalmic procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.