Learn More
Med Vet International 5.5-in. Von Klaus Kelly Forceps, Curved, German Surgical Steel
Catalog No. 502091919
Encompass_Preferred
Kelly forceps are generally used for clamping large blood vessels or manipulating heavy tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1919 Supplier Med Vet International Supplier No. VK1157114
Kelly forceps are generally used for clamping large blood vessels or manipulating heavy tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.