Learn More
Electron Microscopy Sciences Ergonomic Paddle Handle Forceps Curved Serrated Tips, 178 mm
SDP
Catalog No. 501927196
Encompass_Preferred
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles. 178 mm versions are for those preferring a longer forcep and a wider variety of tip choices. Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
OAL: 178 mm
Tip: 1.5 mm
Catalog No. 50-192-7196 Supplier Electron Microscopy Sciences Supplier No. 7793754
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles. 178 mm versions are for those preferring a longer forcep and a wider variety of tip choices. Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
OAL: 178 mm
Tip: 1.5 mm