Learn More
Fine Science Tools Hartman Hemostats, Stainless Steel, Straight, 10 cm
SDP
Catalog No. NC9752594
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1mm/ Clamping Length: 18mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9752594 Supplier Fine Science Tools Supplier No. 1300210
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1mm/ Clamping Length: 18mm/ Alloy / Material: Stainless Steel/ Length: 10cm