Learn More
World Precision Instrument Forceps, 11.5cm, Straight, 1x2 Teeth, tying platform
SDP
Catalog No. 502538949
Encompass
  • 11.5 cm (4.5") Long
  • Straight
  • 1 x 2, 0.1 2mm Teeth
  • 5 mm Tying platform
  • Titanium
Catalog No. 50-253-8949 Supplier World Precision Instrument Supplier No. 555048FT
  • 11.5 cm (4.5") Long
  • Straight
  • 1 x 2, 0.1 2mm Teeth
  • 5 mm Tying platform
  • Titanium