Learn More
World Precision Instrument Tissue Forceps, 12.5 cm, 1x2 Teeth, German
SDP
Catalog No. 50822338
Encompass
Tissue forceps with 1x2 teeth have one tooth on one tip and two teeth on the other tip of the tweezers. These tweezers have a fine tip for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, the 1x2 configuration causes less tissue trauma than surgical tweezers with more teeth. These are available with curved or straight tips. The curved tips offer greater visibility of the operating area. They are 12.5cm (5") long and fit easily into your hand for precision work. These are manufactured with surgical grade stainless steel or our premium German steel, both of which are suitable for sterilization by autoclave or chemical sterilant.
Catalog No. 50-822-338 Supplier World Precision Instrument Supplier No. 15918G
Tissue forceps with 1x2 teeth have one tooth on one tip and two teeth on the other tip of the tweezers. These tweezers have a fine tip for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, the 1x2 configuration causes less tissue trauma than surgical tweezers with more teeth. These are available with curved or straight tips. The curved tips offer greater visibility of the operating area. They are 12.5cm (5") long and fit easily into your hand for precision work. These are manufactured with surgical grade stainless steel or our premium German steel, both of which are suitable for sterilization by autoclave or chemical sterilant.