Learn More
Fine Science Tools Tissue Forceps, 1 x 2 Teeth, Stainless Steel, 12 cm
SDP
Catalog No. NC9915158
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 1.7 x 1.2mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9915158 Supplier Fine Science Tools Supplier No. 1102112
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 1.7 x 1.2mm/ Alloy / Material: Stainless Steel/ Length: 12cm