Learn More
World Precision Instrument Tissue Forceps, 15 cm, 1x2 Teeth
SDP
Catalog No. 50822770
Encompass
Tissue forceps with 1x2 teeth have one tooth on one tip and two teeth on the other tip of the tweezers. These tweezers have a fine tip for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, the 1x2 configuration causes less tissue trauma than surgical tweezers with more teeth. These have straight tips and are 15cm (6") long and fit easily into your hand for precision work. These are manufactured with our premium grade German stainless steel, which is suitable for sterilization by autoclave or chemical sterilant. As with all fine surgical instruments, for best results and a long life for the instruments, always clean your surgical forceps thoroughly after each use.
Catalog No. 50-822-770 Supplier World Precision Instrument Supplier No. 501742G
Tissue forceps with 1x2 teeth have one tooth on one tip and two teeth on the other tip of the tweezers. These tweezers have a fine tip for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, the 1x2 configuration causes less tissue trauma than surgical tweezers with more teeth. These have straight tips and are 15cm (6") long and fit easily into your hand for precision work. These are manufactured with our premium grade German stainless steel, which is suitable for sterilization by autoclave or chemical sterilant. As with all fine surgical instruments, for best results and a long life for the instruments, always clean your surgical forceps thoroughly after each use.