Learn More
World Precision Instrument Round Hollow Handled Forceps, 15cm, Angled, 0.15mm Tips
SDP
Catalog No. 50822277
Encompass
Hollowing out the handles of long, round-handled instruments decreases the overall weight. This makes them efficient and non-fatiguing when operating under a microscope. Made from high quality stainless steel, these instruments are an excellent addition to a microsurgical instrument kit.
Catalog No. 50-822-277 Supplier World Precision Instrument Supplier No. 14209
Hollowing out the handles of long, round-handled instruments decreases the overall weight. This makes them efficient and non-fatiguing when operating under a microscope. Made from high quality stainless steel, these instruments are an excellent addition to a microsurgical instrument kit.