Learn More
Fine Science Tools FORCEPS,17CM,CURVED,SERRATED
SDP
Catalog No. NC1816063
Encompass
Taylor ForcepsAlloy / Material:Stainless SteelLength:17cmTip Shape:CurvedTips:SerratedTip Dimensions:1.5 x 1.05mm
This item is not returnable. View return policy
Catalog No. NC1816063 Supplier Fine Science Tools Supplier No. 1101117
Taylor ForcepsAlloy / Material:Stainless SteelLength:17cmTip Shape:CurvedTips:SerratedTip Dimensions:1.5 x 1.05mm