Learn More
Fine Science Tools Standard Pattern Forceps, Stainless Steel, 14.5 cm
SDP
Catalog No. NC9241882
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 3.25 x 1.6mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm
Catalog No. NC9241882 Supplier Fine Science Tools Supplier No. 1100014
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 3.25 x 1.6mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm