Learn More
Fine Science Tools Dumont #5 - Ceramic Coated Forceps, Straight, 0.05 x 0.014 mm, 11 cm
SDP
Catalog No. NC9791564
Encompass
Style: #5/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.014mm/ Alloy / Material: Ceramic Coated Inox/ Length: 11cm
Catalog No. NC9791564 Supplier Fine Science Tools Supplier No. 1125250
Style: #5/ Tip Shape: Straight/ Tips: Biologie/ Tip Dimensions: 0.05 x 0.014mm/ Alloy / Material: Ceramic Coated Inox/ Length: 11cm