Learn More
Electron Microscopy Sciences Ergonomic Paddle Handle Forceps Straight Serrated Tips, 140 mm
SDP
Catalog No. 501927201
Encompass_Preferred
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles. 178 mm versions are for those preferring a longer forcep and a wider variety of tip choices. Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
OAL: 140 mm
Tip: 1.5 mm
Catalog No. 50-192-7201 Supplier Electron Microscopy Sciences Supplier No. 7793759
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles. 178 mm versions are for those preferring a longer forcep and a wider variety of tip choices. Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
OAL: 140 mm
Tip: 1.5 mm