Learn More
Fine Science Tools Tissue Forceps, 2 x 3 Teeth, Stainless Steel, 14.5 cm
SDP
Catalog No. NC9700898
Encompass
Tip Shape: Straight/ Tips: 2x3 Teeth/ Tip Dimensions: 2.9 x 2mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm
Catalog No. NC9700898 Supplier Fine Science Tools Supplier No. 1102214
Tip Shape: Straight/ Tips: 2x3 Teeth/ Tip Dimensions: 2.9 x 2mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm