Learn More
Fine Science Tools Tissue Forceps, 2 x 3 Teeth, Stainless Steel, 15.5 cm
SDP
Catalog No. NC9152338
Encompass
Tip Shape: Straight/ Tips: 2x3 Teeth/ Tip Dimensions: 3.7 x 2mm/ Alloy / Material: Stainless Steel/ Length: 15.5cm
Catalog No. NC9152338 Supplier Fine Science Tools Supplier No. 1102215
Tip Shape: Straight/ Tips: 2x3 Teeth/ Tip Dimensions: 3.7 x 2mm/ Alloy / Material: Stainless Steel/ Length: 15.5cm