Learn More
World Precision Instrument Dressing Forceps, 30.5cm
SDP
Catalog No. 50822473
Encompass
Dressing Forceps, also known as thumb forceps or pickups, are surgical instruments characterized by their long, thin handles and serrated tips. They are typically used when dressing wounds where they hold gauze and other dressings. They may also be used during wound debridement to remove infected or necrotic tissue or debris from the wound. They may be used for suturing when needle holders are not accessible. Dressing forceps are designed to grip delicate tissue without causing damage, and they are often used in situations where precision and control are important. These dressing forceps have straight blades and rounded (blunt) tips. They are available in various sizes: 12.5 cm (5"), 15.5 cm (6.1"), 25.5cm (10"), or 30.5 (12") long.
Catalog No. 50-822-473 Supplier World Precision Instrument Supplier No. 500365
Dressing Forceps, also known as thumb forceps or pickups, are surgical instruments characterized by their long, thin handles and serrated tips. They are typically used when dressing wounds where they hold gauze and other dressings. They may also be used during wound debridement to remove infected or necrotic tissue or debris from the wound. They may be used for suturing when needle holders are not accessible. Dressing forceps are designed to grip delicate tissue without causing damage, and they are often used in situations where precision and control are important. These dressing forceps have straight blades and rounded (blunt) tips. They are available in various sizes: 12.5 cm (5"), 15.5 cm (6.1"), 25.5cm (10"), or 30.5 (12") long.