Learn More
Fine Science Tools Cover Glass Forceps, Stainless Steel, Straight, 10.5 cm
SDP
Catalog No. NC9079871
Encompass
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 4 x 0.6mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm
Catalog No. NC9079871 Supplier Fine Science Tools Supplier No. 1107402
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 4 x 0.6mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm