Learn More
Fine Science Tools Micro-Adson Forceps, Stainless Steel, Straight, 12 cm
SDP
Catalog No. NC9763233
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.8 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9763233 Supplier Fine Science Tools Supplier No. 1101912
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.8 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 12cm