Learn More
Fine Science Tools Iris Forceps
SDP
Catalog No. NC9206525
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.6 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 7cm
Catalog No. NC9206525 Supplier Fine Science Tools Supplier No. 1106507
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.6 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 7cm