Learn More
Fine Science Tools FORCEPS
SDP
Catalog No. NC9082916
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9082916 Supplier Fine Science Tools Supplier No. 1115110
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm