Learn More
Roboz Surgical FORCEPS
SDP
Supplier:  Roboz Surgical RS4950
Encompass
Dumont Tweezers; Pattern #PP; Heavy Duty; .10 X .06mm Tip Size; Stainless Steel; 135mm Length; Material: Inox; Biologie Tips
Catalog No. NC9787084