Learn More
Electron Microscopy Sciences Thumb Dressing Broad Grip Forceps with 2 mm Smooth Tips, 152 mm
SDP
Catalog No. NC1518560
Encompass_Preferred
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles. The forceps are made of premium grade, hardened stainless steel with a protective coating for extra durability and protection from rust and corrosion. The satin-matt finish reduces eye-fatiguing glare.
Tips: 2 mm Smooth Tips
OAL: 152 mm
Catalog No. NC1518560 Supplier Electron Microscopy Sciences Supplier No. 7793724
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles. The forceps are made of premium grade, hardened stainless steel with a protective coating for extra durability and protection from rust and corrosion. The satin-matt finish reduces eye-fatiguing glare.
Tips: 2 mm Smooth Tips
OAL: 152 mm