Learn More
Med Vet International Miltex Meisterhand Adair Tenaculum Forceps, 6.75in, German, Mh16-51
Catalog No. 502831211
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Adair Tenaculum Forceps. Adair Tenaculum Forceps are commonly used in surgery to secure blood vessels or to steady the cervix and uterus. They are made from premium-grade German stainless steel. Specifications: 6.75in, MH16-51
Catalog No. 50-283-1211 Supplier Med Vet International Supplier No. MILMH1651
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Adair Tenaculum Forceps. Adair Tenaculum Forceps are commonly used in surgery to secure blood vessels or to steady the cervix and uterus. They are made from premium-grade German stainless steel. Specifications: 6.75in, MH16-51