Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, 1x2 Teeth
SDP
Catalog No. 50822366
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-366 Supplier World Precision Instrument Supplier No. 500092
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.