Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, Serrated
SDP
Catalog No. 50822302
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-302 Supplier World Precision Instrument Supplier No. 14226
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.