Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, Serrated, German
SDP
Catalog No. 50822303
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-303 Supplier World Precision Instrument Supplier No. 14226G
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.