Learn More
Med Vet International Miltex Adson Hemostatic Forceps, 7.5in, Straight, German, 7-240
Catalog No. 502832384
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Adson Hemostatic Forceps. Adson Hemostatic Forceps are primarily used for clamping small vessels to help halt blood flow. They are made from premium-grade German stainless steel. Specifications: 7.5in, Straight, 7-240
Catalog No. 50-283-2384 Supplier Med Vet International Supplier No. MIL7240
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Adson Hemostatic Forceps. Adson Hemostatic Forceps are primarily used for clamping small vessels to help halt blood flow. They are made from premium-grade German stainless steel. Specifications: 7.5in, Straight, 7-240