Learn More
World Precision Instrument Adson-Brown Forceps, 12cm, Straight, TC Jaws, 7x7 Teeth
SDP
Catalog No. 50822375
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
  • 12 cm (4.75") Long
  • Tungsten carbide jaws
  • 7x7 Side grasping teeth
  • Stainless steel
This item is not returnable. View return policy
Catalog No. 50-822-375 Supplier World Precision Instrument Supplier No. 500177
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
  • 12 cm (4.75") Long
  • Tungsten carbide jaws
  • 7x7 Side grasping teeth
  • Stainless steel