Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, 1x2 Teeth, German, TC inserts
SDP
Catalog No. 50822378
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-378 Supplier World Precision Instrument Supplier No. 500178G
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.