Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, Serrated, TC Jaws
SDP
Catalog No. 50822391
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-391 Supplier World Precision Instrument Supplier No. 500222
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.