Learn More
Accuflow Systems Saint-Gobain Forceps, PTFE Coated, Nickel-Plated Steel, 115 mm
Catalog No. NC2327707
Encompass_Preferred
PTFE-coated forceps designed for laboratory handling of hot or chemically active materials. Constructed from nickel-plated steel with a heavy-duty PTFE (Teflon) coating, these approximately 115 mm forceps offer a non-stick, chemically inert surface and blunt tips for secure, low-damage grasping.
  • PTFE-coated surface provides a non-stick, chemically inert barrier.
  • Nickel-plated steel body for strength and corrosion resistance.
  • Approx. 115 mm length (114-116 mm reported across listings).
  • Blunt tip style for secure handling without piercing.
  • Suitable for handling hot or chemically active materials.
Catalog No. NC2327707 Supplier Accuflow Systems Supplier No. D1069288
PTFE-coated forceps designed for laboratory handling of hot or chemically active materials. Constructed from nickel-plated steel with a heavy-duty PTFE (Teflon) coating, these approximately 115 mm forceps offer a non-stick, chemically inert surface and blunt tips for secure, low-damage grasping.
  • PTFE-coated surface provides a non-stick, chemically inert barrier.
  • Nickel-plated steel body for strength and corrosion resistance.
  • Approx. 115 mm length (114-116 mm reported across listings).
  • Blunt tip style for secure handling without piercing.
  • Suitable for handling hot or chemically active materials.