Learn More
World Precision Instrument Applying Forceps for Approximator and Vessel Clips, 10cm
SDP
Catalog No. 50822271
Encompass
  • 10 cm (4") Long
  • Use with Vessel Clips 14120, 15911, and 14121
  • Use with the 14084 Approximator
  • Stainless steel
Catalog No. 50-822-271 Supplier World Precision Instrument Supplier No. 14189
  • 10 cm (4") Long
  • Use with Vessel Clips 14120, 15911, and 14121
  • Use with the 14084 Approximator
  • Stainless steel