Learn More
Med Vet International Miltex Vantage Babcock Tissue Hemostatic Forceps, 6.25in, 8mm Wide Jaws
Catalog No. 502522281
Encompass_Preferred
The Miltex Vantage V916-44 Babcock tissue forceps are made from high quality Japanese stainless steel and measure 6.25" in length. They have a finger ring handle with a ratchet lock and 8mm wide fenestrated triangular jaws.
Catalog No. 50-252-2281 Supplier Med Vet International Supplier No. MILV91644
The Miltex Vantage V916-44 Babcock tissue forceps are made from high quality Japanese stainless steel and measure 6.25" in length. They have a finger ring handle with a ratchet lock and 8mm wide fenestrated triangular jaws.