Learn More
World Precision Instrument Forceps Basic Kit Student Grade
SDP
Catalog No. 502538383
Encompass
WPI's Basic Forceps Kits include either three or five popular forceps in an economy grade. These kits are perfect for students, giving them a variety of tip shapes and designs so they can develop proficiency. They are all constructed of stainless steel and may be sterilized. The first kit (WP00118) includes #4, #5, and #7  tweezers. The large kit (WPI00218) includes #4, #5, #7, #1, and #3C. The second kit also includes a canvas pouch. Buying kits lets you save money, too!
Catalog No. 50-253-8383 Supplier World Precision Instrument Supplier No. WP00118
WPI's Basic Forceps Kits include either three or five popular forceps in an economy grade. These kits are perfect for students, giving them a variety of tip shapes and designs so they can develop proficiency. They are all constructed of stainless steel and may be sterilized. The first kit (WP00118) includes #4, #5, and #7  tweezers. The large kit (WPI00218) includes #4, #5, #7, #1, and #3C. The second kit also includes a canvas pouch. Buying kits lets you save money, too!