Learn More
Med Vet International Miltex Vantage Bishop-Harmon Tissue Forceps, 3.5in, with 1x2 Teeth
Catalog No. 502522276
Encompass_Preferred
The Miltex Vantage V918-868 Bishop-Harmon tissue forceps are made from high quality Japanese stainless steel and measure 3.5" in length. They have a non-locking, fenestrated thumb handle and straight tips with 1 x 2 teeth.
Catalog No. 50-252-2276 Supplier Med Vet International Supplier No. MILV918868
The Miltex Vantage V918-868 Bishop-Harmon tissue forceps are made from high quality Japanese stainless steel and measure 3.5" in length. They have a non-locking, fenestrated thumb handle and straight tips with 1 x 2 teeth.