Learn More
Fine Science Tools FORCEPS, BISHOP-HARMON SS, SER
SDP
Catalog No. NC0570187
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm
Catalog No. NC0570187 Supplier Fine Science Tools Supplier No. 1106908
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm