Learn More
Med Vet International Liston Bone Cutting Forceps, 5.5", Angled, German grade, Von Klaus
Catalog No. 503046140
Encompass_Preferred
5.5in. Liston Bone Cutting Forceps is a German grade surgical instrument. They are angled and have a double spring. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6140 Supplier Med Vet International Supplier No. VK1332114
5.5in. Liston Bone Cutting Forceps is a German grade surgical instrument. They are angled and have a double spring. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.