Learn More
Med Vet International Desmarres Chalazion Forceps, 3.5", Large, German Grade, Von Klaus
Catalog No. 503046181
Encompass_Preferred
3.5in. Desmarres Chalazion forceps are German grade surgical instrument use in ophthalmic procedures. They have a large inside ring that is 28mm x 17mm. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6181 Supplier Med Vet International Supplier No. VK1605503
3.5in. Desmarres Chalazion forceps are German grade surgical instrument use in ophthalmic procedures. They have a large inside ring that is 28mm x 17mm. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.