Learn More
World Precision Instrument Crile Forceps, 14cm, Curved, German
SDP
Catalog No. 50822757
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-822-757 Supplier World Precision Instrument Supplier No. 501730G
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."