Learn More
World Precision Instrument Crile Forceps, 14cm, Straight
SDP
Catalog No. 50822591
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-822-591 Supplier World Precision Instrument Supplier No. 501242
 Restricted Item
Add to Cart
Add to Cart
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."