Learn More
World Precision Instrument Crile-Rankin Hemostatic Forceps, 15.9cm, Straight
SDP
Catalog No. 50822725
Encompass
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."
Catalog No. 50-822-725 Supplier World Precision Instrument Supplier No. 501712
Crile hemostatic forceps are atraumatic and non-toothed clamps used to clamp blood vessels, grab tissue or vessels. They are often known as a "snap."